missing translation for 'onlineSavingsMsg'
Learn More

Direct Yellow 26, TCI America™

Catalog No. D147325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D147325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D147325G Supplier TCI America Supplier No. D147325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2829-42-7
Molecular Formula C27H18N6Na2O7
Molecular Weight (g/mol) 584.456
MDL Number MFCD00266524
InChI Key NPQLRSWJRSHFMF-UHFFFAOYSA-L
Synonym Amanil Fast Yellow 5GL, Direct Fast Yellow 5GL, Direct Yellow MC
PubChem CID 73557454
IUPAC Name disodium;3-[[4-[[4-[2-(3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]phenyl]carbamoylamino]phenyl]hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
SMILES C1=CC(=CC=C1NC(=O)NC2=CC=C(C=C2)NN=C3C=CC(=O)C(=C3)C(=O)[O-])NN=C4C=CC(=O)C(=C4)C(=O)[O-].[Na+].[Na+]

Specifications

Quantity 25 g
Formula Weight 584.46
Physical Form Crystalline Powder
Chemical Name or Material Direct Yellow 26