Learn More
Brady™ Direct Thermal Printable Labels
For use in warehouse/industrial ID, lab ID, government asset tracking. Brady™ Direct Thermal Printable Labels are made of topcoated paper material.
Supplier:  Brady™ DT258161
 View more versions of this product
Catalog No. 19-109-050
Only null left
Description
Description
  • Warehouse/Industrial ID, general Lab ID and Government asset tracking applications
  • Made of B-813 and B-816 topcoated paper material
  • B-816: Removable acrylic adhesive
  • B-813: Permamnent acrylic adhesive
Specifications
Specifications
REACH Compliant, RoHS Compliant
White
Direct Thermal Paper (B-816)
Short-term identification
Rectangle
0.0039 in.
Acrylic, Removable
Roll
Direct Thermal Printable Label
BBP™81 Label Printer, BRADY IP Series, BradyPrinter PR Plus Series Printer
6 x 4 in. (W x H)
Product Suggestions
Product Suggestions