missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Direct Red 80

Catalog No. AAB2169306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2169306 5 g
AAB2169314 25 g
AAB2169322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2169306 Supplier Thermo Scientific Chemicals Supplier No. B2169306

CAS: 2610-10-8 | C45H26N10Na6O21S6 | 1373.05 g/mol

Direct red 80 is a dye useful as a cardiac muscle stain and as a model compound in adsorption studies. It has also been utilized to compare efficacy of textile dye removal in aqueous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Direct red 80 is a dye useful as a cardiac muscle stain and as a model compound in adsorption studies. It has also been utilized to compare efficacy of textile dye removal in aqueous solutions.

Solubility
Partly soluble in water, very slightly soluble in ethanol, and cellosolve. Insoluble in many organic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 2610-10-8
MDL Number MFCD00054389

Specifications

Molecular Formula C45H26N10Na6O21S6
Solubility Information Partly soluble in water,very slightly soluble in ethanol,and cellosolve. Insoluble in many organic solvents.
InChI Key LDCKXVAPQFUIOI-ZFLDVXHKSA-H
SMILES C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C(=CC4=C(C3=O)C=CC(=C4)NC(=O)NC5=CC6=C(C=C5)C(=O)C(=NNC7=C(C=C(C=C7)N=NC8=CC=C(C=C8)S(=O)(=O)[O-])S(=O)(=O)[O-])C(=C6)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+]
IUPAC Name hexasodium;4-oxo-7-[[5-oxo-7-sulfonato-6-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalen-2-yl]carbamoylamino]-3-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2-sulfonate
Molecular Weight (g/mol) 1373.05
PubChem CID 57369623
Formula Weight 1373.09
Quantity 5 g
Chemical Name or Material Direct Red 80
EINECSNumber 220-027-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only