missing translation for 'onlineSavingsMsg'
Learn More

Direct Blue 2, TCI America™

Catalog No. C052925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C052925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C052925G Supplier TCI America Supplier No. C052925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2429-73-4
Molecular Formula C32H21N6Na3O11S3
Molecular Weight (g/mol) 830.70
MDL Number MFCD00059132
InChI Key ZAOMBAUENFURGO-OWOLPYOXSA-K
Synonym Diazo Black BHN, Chlorazol Black BH
PubChem CID 73557434
IUPAC Name trisodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-7-amino-1-oxo-3-sulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].[Na+].NC1=CC=C2C=C(\C(=N/NC3=CC=C(C=C3)C3=CC=C(N\N=C4\C(=O)C5=C(N)C=C(C=C5C=C4S([O-])(=O)=O)S([O-])(=O)=O)C=C3)C(=O)C2=C1)S([O-])(=O)=O

Specifications

Quantity 25 g
Formula Weight 830.70
Physical Form Crystalline Powder
Chemical Name or Material Direct Blue 2
EINECSNumber (5)-1282
RTECSNumber QJ6158000
TSCA Yes