accessibility menu, dialog, popup

UserName

CAS RN 2429-73-4

IUPAC Name: trisodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-7-amino-1-oxo-3-sulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: trisodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-7-amino-1-oxo-3-sulfonatonaphthalen-2-ylidene]hydrazin-1-yl}-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 830.7
Molecular Formula: C32H21N6Na3O11S3
InChi Key: ZAOMBAUENFURGO-OWOLPYOXSA-K
SMILES: [Na+].[Na+].[Na+].NC1=CC=C2C=C(\C(=N/NC3=CC=C(C=C3)C3=CC=C(N\N=C4\C(=O)C5=C(N)C=C(C=C5C=C4S([O-])(=O)=O)S([O-])(=O)=O)C=C3)C(=O)C2=C1)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results