missing translation for 'onlineSavingsMsg'
Learn More

Dipyrido[3,2-a:2',3'-c]phenazine 98.0+%, TCI America™

Catalog No. p-7128249
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4379200MG 200 mg
D43791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4379200MG Supplier TCI America Supplier No. D4379200MG

Chemical Identifiers

CAS 19535-47-8
Molecular Formula C18H10N4
Molecular Weight (g/mol) 282.306
MDL Number MFCD04113602
InChI Key BVQAWSJMUYMNQN-UHFFFAOYSA-N
Synonym Dppz
PubChem CID 3081616
ChEBI CHEBI:30622
IUPAC Name quinoxalino[2,3-f][1,10]phenanthroline
SMILES C1=CC=C2C(=C1)N=C3C4=C(C5=C(C3=N2)C=CC=N5)N=CC=C4

Specifications

Melting Point 248°C
Quantity 200 mg
Formula Weight 282.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dipyrido[3,2-a:2′,3′-c]phenazine