missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene Glycol (mixture of isomers) 95.0+%, TCI America™
Supplier: TCI America D0933500G
Specifications
| Dipropylene Glycol (mixture of isomers) | |
| Colorless | |
| MFCD00004538 | |
| 134.18 | |
| Liquid |
| 25265-71-8 | |
| C6H14O3 | |
| 500 g | |
| ≥95.0% (GC) |
Chemical Identifiers
| 25265-71-8 | |
| C6H14O3 | |
| MFCD00004538 |
Safety and Handling
EINECSNumber : (2)-0413
RTECSNumber : UB8765000
TSCA : Yes