missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene Glycol (mixture of isomers) 95.0+%, TCI America™

Catalog No. p-7126509
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
D093325G 25 g
D0933500G 500 g
2 options

Catalog No. D093325G

Supplier: TCI America D093325G

Only null left

Chemical Identifiers

CAS 25265-71-8
Molecular Formula C6H14O3
MDL Number MFCD00004538

Specifications

Color Colorless
Quantity 25 g
Formula Weight 134.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropylene Glycol (mixture of isomers)
EINECSNumber (2)-0413
RTECSNumber UB8765000
TSCA Yes