missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene Glycol Methyl Ether Acetate, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor D11114LTGL
Specifications
| 0.0005 | |
| 1 | |
| C9H18O4 | |
| USDXBNSCKZJDLC-UHFFFAOYNA-N | |
| 1-(3-methoxypropoxy)propyl acetate | |
| 0.98 |
| 88917-22-0 | |
| Amber Glass Bottle | |
| 4 L | |
| CCC(OCCCOC)OC(C)=O | |
| 190.24 | |
| Reagent |
Chemical Identifiers
| 88917-22-0 | |
| 190.24 | |
| 1-(3-methoxypropoxy)propyl acetate |
| C9H18O4 | |
| USDXBNSCKZJDLC-UHFFFAOYNA-N | |
| CCC(OCCCOC)OC(C)=O |
CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.