missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene Glycol Methyl Ether Acetate, 98%, Spectrum™ Chemical

Catalog No. p-7165470
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610294 250 mL Amber Glass Bottle
18610295 1 L Amber Glass Bottle
18610296 4 L Amber Glass Bottle
18610297 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610294 Supplier Spectrum Chemical Mfg Cor Supplier No. D1111250ML
Only null left

Chemical Identifiers

CAS 88917-22-0
Molecular Formula C9H18O4
Molecular Weight (g/mol) 190.24
InChI Key USDXBNSCKZJDLC-UHFFFAOYNA-N
IUPAC Name 1-(3-methoxypropoxy)propyl acetate
SMILES CCC(OCCCOC)OC(C)=O

Specifications

Acidity 0.0005
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 250 mL
Percent Purity 0.98
Grade Reagent

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.