missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene Glycol Methyl Ether, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor D11101LT
Specifications
| 0.0001 | |
| 1 | |
| C7H16O3 | |
| 0.945 - 0.957 | |
| COC(C)COC(C)CO | |
| 148.20 | |
| Reagent |
| 34590-94-8 | |
| Amber Glass Bottle | |
| 1 L | |
| CUDYYMUUJHLCGZ-UHFFFAOYNA-N | |
| 2-(2-methoxypropoxy)propan-1-ol | |
| 0.99 |
Chemical Identifiers
| 34590-94-8 | |
| 148.20 | |
| 2-(2-methoxypropoxy)propan-1-ol |
| C7H16O3 | |
| CUDYYMUUJHLCGZ-UHFFFAOYNA-N | |
| COC(C)COC(C)CO |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.