missing translation for 'onlineSavingsMsg'
Learn More

Dipropylene Glycol Methyl Ether, 99%, Spectrum™ Chemical

Catalog No. 18610290
Change view
Click to view available options
Quantity:
25 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610290 25 mL Amber Glass Bottle
18610291 1 L Amber Glass Bottle
18610292 4 L Amber Glass Bottle
18610293 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610290 Supplier Spectrum Chemical Mfg Cor Supplier No. D111025ML

Chemical Identifiers

CAS 34590-94-8
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.20
InChI Key CUDYYMUUJHLCGZ-UHFFFAOYNA-N
IUPAC Name 2-(2-methoxypropoxy)propan-1-ol
SMILES COC(C)COC(C)CO

Specifications

Acidity 0.0001
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 mL
Specific Gravity 0.945 - 0.957
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.