missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene glycol, 99%, mixture of isomers
CAS: 25265-71-8 | C6H14O3 | 134.18 g/mol
$167.12 - $280.00
Chemical Identifiers
| CAS | 25265-71-8 |
|---|---|
| Molecular Formula | C6H14O3 |
| Molecular Weight (g/mol) | 134.18 |
| MDL Number | MFCD00051023 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC409150010
|
Thermo Scientific Chemicals
409150010 |
1 kg | Glass Bottle |
Each for $167.12
|
|
||||
|
AC409150030
|
Thermo Scientific Chemicals
409150030 |
3 kg | Plastic drum |
Each for $280.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Note: The chemical structure above shows only one of the molecules included in this mixture.Chemical Identifiers
| 25265-71-8 | |
| C6H14O3 | |
| 134.18 | |
| MFCD00051023 |
Specifications
| 25265-71-8 | |
| 1.0200g/mL | |
| 138°C | |
| 98.5% min. (GC) | |
| C6H14O3 | |
| 1.4400 to 1.4420 | |
| 1 kg | |
| Solubility in water: miscible. Other solubilities: soluble in ethanol | |
| 134.18 | |
| Liquid |
| Orange-Red to Yellow | |
| 229.0°C to 232.0°C | |
| Authentic | |
| Glass Bottle | |
| CH3CH(OH)CH2OCH2CH(OH)CH3 | |
| MFCD00051023 | |
| 1.02 | |
| 134.18 | |
| 99% | |
| Dipropylene glycol, 99% |
Safety and Handling
EINECSNumber : 246-770-3
RTECSNumber : UB8765000
TSCA : TSCA
RUO – Research Use Only