missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropylene glycol, 99%, mixture of isomers
CAS: 25265-71-8 | C6H14O3 | 134.18 g/mol
Supplier: Thermo Scientific Chemicals 409150030
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Note: The chemical structure above shows only one of the molecules included in this mixture.Specifications
| Dipropylene glycol | |
| 25265-71-8 | |
| 1.0200g/mL | |
| 138°C | |
| 98.5% min. (GC) | |
| C6H14O3 | |
| 1.4400 to 1.4420 | |
| 3 kg | |
| Solubility in water: miscible. Other solubilities: soluble in ethanol | |
| 134.18 | |
| Liquid |
| 99% | |
| Orange-Red to Yellow | |
| 229.0°C to 232.0°C | |
| Authentic | |
| Plastic drum | |
| CH3CH(OH)CH2OCH2CH(OH)CH3 | |
| MFCD00051023 | |
| 1.02 | |
| 134.18 | |
| 99% |
Chemical Identifiers
| 25265-71-8 | |
| C6H14O3 | |
| 134.18 | |
| MFCD00051023 |
Safety and Handling
EINECSNumber : 246-770-3
RTECSNumber : UB8765000
TSCA : TSCA
RUO – Research Use Only