missing translation for 'onlineSavingsMsg'
Learn More

Dipropylammonium Acetate (ca. 0.5mol/L in Water) [Ion-Pair Reagent for LC-MS], TCI America™

Catalog No. A570310ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A570310ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A570310ML Supplier TCI America Supplier No. A570310ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 114389-69-4
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.25
MDL Number MFCD03093617
InChI Key LKDMDHMCNGRIAS-UHFFFAOYSA-N
Synonym IPC-DPAA
PubChem CID 16218734
IUPAC Name acetic acid; dipropylamine
SMILES CC(O)=O.CCCNCCC

Specifications

Color Colorless
Quantity 10 mL
Formula Weight 161.25
Physical Form Liquid
Chemical Name or Material Dipropylammonium Acetate (ca. 0.5mol/L in Water) [Ion-Pair Reagent for LC-MS]