accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 114389-69-4

H3CNHCH3CH3OHO

CAS RN 114389-69-4

IUPAC Name: acetic acid; dipropylamine
Synonyms: acetic acid; dipropylamine
Molecular Weight (g/mol): 161.25
Molecular Formula: C8H19NO2
InChi Key: LKDMDHMCNGRIAS-UHFFFAOYSA-N
SMILES: CC(O)=O.CCCNCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.