missing translation for 'onlineSavingsMsg'
Learn More

Dipropyl Sulfone 99.0+%, TCI America™

Catalog No. D117125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D117125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D117125G Supplier TCI America Supplier No. D117125G
Only null left

Chemical Identifiers

CAS 598-03-8
Molecular Formula C6H14O2S
Molecular Weight (g/mol) 150.236
MDL Number MFCD00007570
InChI Key JEXYCADTAFPULN-UHFFFAOYSA-N
Synonym propyl sulfone, dipropyl sulfone, di-n-propyl sulfone, propane, 1,1'-sulfonylbis, dipropyl sulphone, n-propyl sulfone, unii-581dro3v26, dipropylsulfone, di-n-propylsulfone, propylsulfonyl propane
PubChem CID 11709
IUPAC Name 1-propylsulfonylpropane
SMILES CCCS(=O)(=O)CCC

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 150.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dipropyl Sulfone