accessibility menu, dialog, popup

UserName

CAS RN 598-03-8

CH3SOOH3C

CAS RN 598-03-8

IUPAC Name: 1-(propane-1-sulfonyl)propane
Synonyms: propyl sulfone
Molecular Weight (g/mol): 150.24
Molecular Formula: C6H14O2S
InChi Key: JEXYCADTAFPULN-UHFFFAOYSA-N
SMILES: CCCS(=O)(=O)CCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results