missing translation for 'onlineSavingsMsg'
Learn More

Dipropyl Sulfate 90.0+%, TCI America™

Catalog No. D163825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D163825G 25 g
D1638500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D163825G Supplier TCI America Supplier No. D163825G
Only null left

Chemical Identifiers

CAS 598-05-0
Molecular Formula C6H14O4S
Molecular Weight (g/mol) 182.234
MDL Number MFCD00059243
InChI Key JYCKNDWZDXGNBW-UHFFFAOYSA-N
PubChem CID 69015
IUPAC Name dipropyl sulfate
SMILES CCCOS(=O)(=O)OCCC

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
UN Number 2922
Formula Weight 182.23
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropyl Sulfate [Alkylating Agent]
EINECSNumber (2)-1673
TSCA Yes