accessibility menu, dialog, popup

UserName

CH3OSOOOH3C

CAS RN 598-05-0

IUPAC Name: dipropyl sulfate
Synonyms: dipropyl sulfate
Molecular Weight (g/mol): 182.23
Molecular Formula: C6H14O4S
InChi Key: JYCKNDWZDXGNBW-UHFFFAOYSA-N
SMILES: CCCOS(=O)(=O)OCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results