missing translation for 'onlineSavingsMsg'
Learn More

Dipropyl Phthalate 98.0+%, TCI America™

Catalog No. P030625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P030625ML 25 mL
P0306100ML 100 mL
P0306500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P030625ML Supplier TCI America Supplier No. P030625ML

Chemical Identifiers

CAS 131-16-8
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.294
MDL Number MFCD00009371
InChI Key MQHNKCZKNAJROC-UHFFFAOYSA-N
Synonym dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss
PubChem CID 8559
ChEBI CHEBI:60069
IUPAC Name dipropyl benzene-1,2-dicarboxylate
SMILES CCCOC(=O)C1=CC=CC=C1C(=O)OCCC

Specifications

Color Yellow
Boiling Point 305°C
Quantity 25 mL
UN Number 3082
Formula Weight 250.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropyl Phthalate
RTECSNumber TI1940000
TSCA Yes