missing translation for 'onlineSavingsMsg'
Learn More

Dipropyl phthalate, 99+%

Catalog No. AC409160250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409160250 25 g Glass bottle
AC409161000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC409160250 Supplier Thermo Scientific Chemicals Supplier No. 409160250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 131-16-8 | C14H18O4 | 250.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 131-16-8
InChI Key MQHNKCZKNAJROC-UHFFFAOYSA-N
Synonym dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss
PubChem CID 8559
ChEBI CHEBI:60069
IUPAC Name dipropyl benzene-1,2-dicarboxylate
SMILES CCCOC(=O)C1=CC=CC=C1C(=O)OCCC

Specifications

Color Colorless
Density 1.0780g/mL
Boiling Point 317.5°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C14H18O4
Refractive Index 1.4960 to 1.4980
Quantity 25 g
Specific Gravity 1.078
Solubility Information Solubility in water: in water: 0.015% by weight.
Molecular Weight (g/mol) 250.29
Formula Weight 250.29
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dipropyl phthalate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-015-8

RUO – Research Use Only