missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropyl 2,5-Pyridinedicarboxylate 98.0+%, TCI America™
Supplier: TCI America P083525G
Specifications
| Dipropyl 2,5-Pyridinedicarboxylate | |
| Yellow | |
| C13H17NO4 | |
| 25 g | |
| IITCWRFYJWUUPC-UHFFFAOYSA-N | |
| dipropyl pyridine-2,5-dicarboxylate | |
| 8693 | |
| ≥98.0% (GC) |
| 136-45-8 | |
| 175°C | |
| MFCD00055315 | |
| dipropyl 2,5-pyridinedicarboxylate, dipropyl isocinchomeronate, di-propylisocinchomeronate, repper 333, di-n-propyl isocinchomeronate, 2,5-pyridinedicarboxylic acid, dipropyl ester, mgk r-326, caswell no. 400, isocinchomeronic acid, dipropyl ester, mgk repellent 326 | |
| CCCOC(=O)C1=CN=C(C=C1)C(=O)OCCC | |
| 251.282 | |
| 251.28 | |
| Liquid |
Chemical Identifiers
| 136-45-8 | |
| 251.282 | |
| IITCWRFYJWUUPC-UHFFFAOYSA-N | |
| 8693 | |
| CCCOC(=O)C1=CN=C(C=C1)C(=O)OCCC |
| C13H17NO4 | |
| MFCD00055315 | |
| dipropyl 2,5-pyridinedicarboxylate, dipropyl isocinchomeronate, di-propylisocinchomeronate, repper 333, di-n-propyl isocinchomeronate, 2,5-pyridinedicarboxylic acid, dipropyl ester, mgk r-326, caswell no. 400, isocinchomeronic acid, dipropyl ester, mgk repellent 326 | |
| dipropyl pyridine-2,5-dicarboxylate |
Safety and Handling
EINECSNumber : (5)-0746
RTECSNumber : US8000000
TSCA : No