missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipropyl 2,5-Pyridinedicarboxylate 98.0+%, TCI America™
$34.66 - $96.26
Chemical Identifiers
| CAS | 136-45-8 |
|---|---|
| Molecular Formula | C13H17NO4 |
| Molecular Weight (g/mol) | 251.282 |
| MDL Number | MFCD00055315 |
| InChI Key | IITCWRFYJWUUPC-UHFFFAOYSA-N |
| Synonym | dipropyl 2,5-pyridinedicarboxylate, dipropyl isocinchomeronate, di-propylisocinchomeronate, repper 333, di-n-propyl isocinchomeronate, 2,5-pyridinedicarboxylic acid, dipropyl ester, mgk r-326, caswell no. 400, isocinchomeronic acid, dipropyl ester, mgk repellent 326 |
| PubChem CID | 8693 |
| IUPAC Name | dipropyl pyridine-2,5-dicarboxylate |
| SMILES | CCCOC(=O)C1=CN=C(C=C1)C(=O)OCCC |
Chemical Identifiers
| 136-45-8 | |
| 251.282 | |
| IITCWRFYJWUUPC-UHFFFAOYSA-N | |
| 8693 | |
| CCCOC(=O)C1=CN=C(C=C1)C(=O)OCCC |
| C13H17NO4 | |
| MFCD00055315 | |
| dipropyl 2,5-pyridinedicarboxylate, dipropyl isocinchomeronate, di-propylisocinchomeronate, repper 333, di-n-propyl isocinchomeronate, 2,5-pyridinedicarboxylic acid, dipropyl ester, mgk r-326, caswell no. 400, isocinchomeronic acid, dipropyl ester, mgk repellent 326 | |
| dipropyl pyridine-2,5-dicarboxylate |
Specifications
| 136-45-8 | |
| 175°C | |
| MFCD00055315 | |
| dipropyl 2,5-pyridinedicarboxylate, dipropyl isocinchomeronate, di-propylisocinchomeronate, repper 333, di-n-propyl isocinchomeronate, 2,5-pyridinedicarboxylic acid, dipropyl ester, mgk r-326, caswell no. 400, isocinchomeronic acid, dipropyl ester, mgk repellent 326 | |
| CCCOC(=O)C1=CN=C(C=C1)C(=O)OCCC | |
| 251.282 | |
| 251.28 | |
| Liquid |
| Yellow | |
| C13H17NO4 | |
| 5 g | |
| IITCWRFYJWUUPC-UHFFFAOYSA-N | |
| dipropyl pyridine-2,5-dicarboxylate | |
| 8693 | |
| ≥98.0% (GC) | |
| Dipropyl 2,5-Pyridinedicarboxylate |
Safety and Handling
EINECSNumber : (5)-0746
RTECSNumber : US8000000
TSCA : No