missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipotassium Anthraquinone-1,8-disulfonate 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 444.51 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dipotassium Anthraquinone-1,8-disulfonate |
Safety and Handling
| EINECSNumber | (4)-0725 |
| RTECSNumber | CB0554500 |
| TSCA | No |