accessibility menu, dialog, popup

UserName

CAS RN 14938-42-2

KKOSOOOOSOOO

CAS RN 14938-42-2

IUPAC Name: dipotassium 9,10-dioxo-9,10-dihydroanthracene-1,8-disulfonate
Synonyms: dipotassium 9,10-dioxoanthracene-1,8-disulfonate
Molecular Weight (g/mol): 444.51
Molecular Formula: C14H6K2O8S2
InChi Key: PWPFDCNGQQSVJK-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-]S(=O)(=O)C1=CC=CC2=C1C(=O)C1=C(C=CC=C1C2=O)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results