missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipivaloylmethane 97.0+%, TCI America™
$105.89 - $300.33
Chemical Identifiers
| CAS | 1118-71-4 |
|---|---|
| Molecular Formula | C11H20O2 |
| Molecular Weight (g/mol) | 184.279 |
| MDL Number | MFCD00008848 |
| InChI Key | YRAJNWYBUCUFBD-UHFFFAOYSA-N |
| Synonym | dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, pubchem12497 |
| PubChem CID | 70700 |
| IUPAC Name | 2,2,6,6-tetramethylheptane-3,5-dione |
| SMILES | CC(C)(C)C(=O)CC(=O)C(C)(C)C |
Chemical Identifiers
| 1118-71-4 | |
| 184.279 | |
| YRAJNWYBUCUFBD-UHFFFAOYSA-N | |
| 70700 | |
| CC(C)(C)C(=O)CC(=O)C(C)(C)C |
| C11H20O2 | |
| MFCD00008848 | |
| dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, pubchem12497 | |
| 2,2,6,6-tetramethylheptane-3,5-dione |
Specifications
| 1118-71-4 | |
| 80°C | |
| MFCD00008848 | |
| dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, pubchem12497 | |
| CC(C)(C)C(=O)CC(=O)C(C)(C)C | |
| 184.279 | |
| 184.28 | |
| Liquid |
| Yellow | |
| C11H20O2 | |
| 5 g | |
| YRAJNWYBUCUFBD-UHFFFAOYSA-N | |
| 2,2,6,6-tetramethylheptane-3,5-dione | |
| 70700 | |
| ≥97.0% (GC) | |
| Dipivaloylmethane |
Safety and Handling
TSCA : Yes