missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylsilanediol 98.0+%, TCI America™
Supplier: TCI America D231625G
Specifications
| Diphenylsilanediol | |
| 147°C | |
| C12H12O2Si | |
| 25 g | |
| diphenylsilanediol, silanediol, diphenyl, diphenyldihydroxysilane, difenyl-dihydroxysilan, silane, dihydroxydiphenyl, dihydroxy diphenyl silane, diphenylsilandiol, silanediol, 1,1-diphenyl, unii-36x37p8nbb, difenyl-dihydroxysilan czech | |
| C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(O)O | |
| 216.311 | |
| 216.31 | |
| Crystalline Powder |
| 947-42-2 | |
| White | |
| MFCD00002101 | |
| 3077 | |
| OLLFKUHHDPMQFR-UHFFFAOYSA-N | |
| dihydroxy(diphenyl)silane | |
| 13693 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 947-42-2 | |
| 216.311 | |
| OLLFKUHHDPMQFR-UHFFFAOYSA-N | |
| 13693 | |
| C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(O)O |
| C12H12O2Si | |
| MFCD00002101 | |
| diphenylsilanediol, silanediol, diphenyl, diphenyldihydroxysilane, difenyl-dihydroxysilan, silane, dihydroxydiphenyl, dihydroxy diphenyl silane, diphenylsilandiol, silanediol, 1,1-diphenyl, unii-36x37p8nbb, difenyl-dihydroxysilan czech | |
| dihydroxy(diphenyl)silane |
Safety and Handling
EINECSNumber : (3)-2638
RTECSNumber : VV3640000
TSCA : Yes