missing translation for 'onlineSavingsMsg'
Learn More

Diphenylsilanediol, 98%

Catalog No. AAA1049236
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1049236 500 g
AAA1049222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1049236 Supplier Thermo Scientific Chemicals Supplier No. A1049236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 947-42-2 | C12H12O2Si | 216.311 g/mol

Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.

Solubility
Reacts with water.

Notes
Stable under normal temperatures and pressures. Incompatible with Oxidizing agents. Store at room temperature. Air & Light Sensitive.

Chemical Identifiers

CAS 947-42-2
Molecular Formula C12H12O2Si
Molecular Weight (g/mol) 216.311
MDL Number MFCD00002101
InChI Key OLLFKUHHDPMQFR-UHFFFAOYSA-N
Synonym diphenylsilanediol, silanediol, diphenyl, diphenyldihydroxysilane, difenyl-dihydroxysilan, silane, dihydroxydiphenyl, dihydroxy diphenyl silane, diphenylsilandiol, silanediol, 1,1-diphenyl, unii-36x37p8nbb, difenyl-dihydroxysilan czech
PubChem CID 13693
IUPAC Name dihydroxy(diphenyl)silane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(O)O

Specifications

Melting Point ∼150°C (decomposition)
Quantity 500 g
Odor Odorless
Linear Formula (C6H5Si(OH)2
UN Number UN1325
Beilstein 2523445
Formula Weight 216.31
Percent Purity 98%
Chemical Name or Material Diphenylsilanediol
Hazard Category Warning
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement GHS P Statement
P210
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
RTECSNumber VV3640000
TSCA TSCA

RUO – Research Use Only