missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphoryl Azide 97.0+%, TCI America™

Catalog No. p-7126867
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16725G 5 g
D167225G 25 g
D1672250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D16725G Supplier TCI America Supplier No. D16725G
Only null left

Chemical Identifiers

CAS 26386-88-9
Molecular Formula C12H10N3O3P
Molecular Weight (g/mol) 275.20
MDL Number MFCD00001987
InChI Key SORGEQQSQGNZFI-UHFFFAOYSA-N
Synonym diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165av
PubChem CID 123414
IUPAC Name {[azido(phenoxy)phosphoryl]oxy}benzene
SMILES [N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
UN Number 3278
Formula Weight 275.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diphenylphosphoryl Azide
EINECSNumber (3)-3359
TSCA No
Recommended Storage Refrigerator