missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphine Oxide 98.0+%, TCI America™

$272.16 - $933.89

Chemical Identifiers

CAS 4559-70-0
Molecular Formula C12H11OP
Molecular Weight (g/mol) 202.19
MDL Number MFCD00002079
InChI Key ASUOLLHGALPRFK-UHFFFAOYSA-N
Synonym diphenylphosphine oxide, phosphine oxide, diphenyl, diphenyl phosphine oxide, hpoph2, asuollhgalprfk-uhfffaoysa-n, dppo, diphenylphosphane oxide, phenylphosphonoylbenzene, diphenylphosphino-1-one, pubchem23810
PubChem CID 6327869
IUPAC Name (phenylphosphoroso)benzene
SMILES O=P(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D28265G
View Documents
TCI America
D28265G
5 g N/A
Only null left
 
D282625G
View Documents
TCI America
D282625G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4559-70-0
202.19
ASUOLLHGALPRFK-UHFFFAOYSA-N
6327869
O=P(C1=CC=CC=C1)C1=CC=CC=C1
C12H11OP
MFCD00002079
diphenylphosphine oxide, phosphine oxide, diphenyl, diphenyl phosphine oxide, hpoph2, asuollhgalprfk-uhfffaoysa-n, dppo, diphenylphosphane oxide, phenylphosphonoylbenzene, diphenylphosphino-1-one, pubchem23810
(phenylphosphoroso)benzene

Specifications

4559-70-0
105°C
MFCD00002079
diphenylphosphine oxide, phosphine oxide, diphenyl, diphenyl phosphine oxide, hpoph2, asuollhgalprfk-uhfffaoysa-n, dppo, diphenylphosphane oxide, phenylphosphonoylbenzene, diphenylphosphino-1-one, pubchem23810
O=P(C1=CC=CC=C1)C1=CC=CC=C1
202.19
202.19
Crystalline Powder
White-Yellow
C12H11OP
5 g
ASUOLLHGALPRFK-UHFFFAOYSA-N
(phenylphosphoroso)benzene
6327869
≥98.0% (GC)
Diphenylphosphine Oxide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'