accessibility menu, dialog, popup

UserName

CAS RN 4559-70-0

IUPAC Name: (phenylphosphoroso)benzene
Synonyms: diphenylphosphine oxide
Molecular Weight (g/mol): 202.19
Molecular Formula: C12H11OP
InChi Key: ASUOLLHGALPRFK-UHFFFAOYSA-N
SMILES: O=P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)