missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyliodonium Chloride 98.0+%, TCI America™
$300.11 - $1,198.59
Chemical Identifiers
| CAS | 1483-72-3 |
|---|---|
| Molecular Formula | C12H10ClI |
| Molecular Weight (g/mol) | 316.57 |
| MDL Number | MFCD00011909 |
| InChI Key | RSJLWBUYLGJOBD-UHFFFAOYSA-M |
| Synonym | diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9 |
| PubChem CID | 73870 |
| IUPAC Name | diphenyliodanium chloride |
| SMILES | [Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 1483-72-3 | |
| 316.57 | |
| RSJLWBUYLGJOBD-UHFFFAOYSA-M | |
| 73870 | |
| [Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1 |
| C12H10ClI | |
| MFCD00011909 | |
| diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9 | |
| diphenyliodanium chloride |
Specifications
| 1483-72-3 | |
| C12H10ClI | |
| 5 g | |
| diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9 | |
| [Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1 | |
| 316.57 | |
| 316.57 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00011909 | |
| 2811 | |
| RSJLWBUYLGJOBD-UHFFFAOYSA-M | |
| diphenyliodanium chloride | |
| 73870 | |
| ≥98.0% (T) | |
| Diphenyliodonium Chloride |
Safety and Handling
RTECSNumber : NN6651666
TSCA : Yes
Recommended Storage : Refrigerator