missing translation for 'onlineSavingsMsg'
Learn More

Diphenyliodonium Chloride 98.0+%, TCI America™

Catalog No. D23565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23565G 5 g
D235625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23565G Supplier TCI America Supplier No. D23565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1483-72-3
Molecular Formula C12H10ClI
Molecular Weight (g/mol) 316.57
MDL Number MFCD00011909
InChI Key RSJLWBUYLGJOBD-UHFFFAOYSA-M
Synonym diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9
PubChem CID 73870
IUPAC Name diphenyliodanium chloride
SMILES [Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 316.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyliodonium Chloride
RTECSNumber NN6651666
TSCA Yes
Recommended Storage Refrigerator