Learn More
Diphenyliodonium chloride, 98+%
CAS: 1483-72-3 | C12H10ClI | 316.57 g/mol
$187.12 - $2865.10
Chemical Identifiers
| CAS | 1483-72-3 |
|---|---|
| Molecular Formula | C12H10ClI |
| Molecular Weight (g/mol) | 316.57 |
| MDL Number | MFCD00011909 |
| InChI Key | RSJLWBUYLGJOBD-UHFFFAOYSA-M |
| Synonym | diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9 |
| PubChem CID | 73870 |
| IUPAC Name | diphenyliodanium;chloride |
| SMILES | [Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1514909
|
Thermo Scientific Chemicals
A1514909 |
10 g |
Each for $187.12
|
|
|||||
|
AAA1514918
|
Thermo Scientific Chemicals
A1514918 |
50 g |
Each for $785.56
|
|
|||||
|
AAA1514930
|
Thermo Scientific Chemicals
A1514930 |
250 g |
Each for $2,865.10
|
|
|||||
Description
Diphenyliodonium chloride is used as a reagent for electrophilic phenylation . It is used as a reactant for copper-catalyzed cross coupling reactions of purinesband arylation of anilines. It is involved in the preparation of acetylenic arylselenide and (arylmethylene)oxindoles. Further, it is employed in the preparation of aryl alkynes through Sonogashira coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiphenyliodonium chloride is used as a reagent for electrophilic phenylation . It is used as a reactant for copper-catalyzed cross coupling reactions of purinesband arylation of anilines. It is involved in the preparation of acetylenic arylselenide and (arylmethylene)oxindoles. Further, it is employed in the preparation of aryl alkynes through Sonogashira coupling reactions.
Solubility
Soluble in water and methanol.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 1483-72-3 | |
| 316.57 | |
| RSJLWBUYLGJOBD-UHFFFAOYSA-M | |
| 73870 | |
| [Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1 |
| C12H10ClI | |
| MFCD00011909 | |
| diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9 | |
| diphenyliodanium;chloride |
Specifications
| 1483-72-3 | |
| C12H10ClI | |
| 10 g | |
| 3574977 | |
| 14,7464 | |
| Soluble in water and methanol. | |
| [Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1 | |
| 316.57 | |
| 316.57 | |
| Diphenyliodonium chloride |
| ∼230°C (decomposition) | |
| MFCD00011909 | |
| UN2811 | |
| Light sensitive | |
| diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9 | |
| RSJLWBUYLGJOBD-UHFFFAOYSA-M | |
| diphenyliodanium;chloride | |
| 73870 | |
| ≥98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 216-049-8
RTECSNumber : NN6651666
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only