missing translation for 'onlineSavingsMsg'
Learn More

Diphénylcarbazide, Grade ACS, LabChem™

Numéro de catalogue. LC136558
Modifier l'affichage
Click to view available options
Quantité:
25 g
Emballage:
Verre ambré
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
LC136558 25 g Verre ambré
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. LC136558 Fournisseur LabChem Code fournisseur LC136558

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 140-22-7
Formule moléculaire C13H14N4O
Poids moléculaire (g/mol) 242.282
Clé InChI KSPIHGBHKVISFI-UHFFFAOYSA-N
Synonyme 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
PubChem CID 8789
ChEBI CHEBI:4641
Nom de l’IUPAC 1,3-dianilinourea
SOURIRES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2

Spécifications

Nom ou substance chimique Diphenylcarbazide
CAS % maximal 100
Couleur Blanc
Emballage Verre ambré
Formule linéaire C13H14N4O
Quantité 25 g
Poids de formule 242.28
Niveau ACS
Forme physique Poudre
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Température ambiante