missing translation for 'onlineSavingsMsg'
Learn More

Diphenylcarbamyl chloride, 98%

CAS: 83-01-2 | C13H10ClNO | 231.68 g/mol

$127.67 - $288.76

Chemical Identifiers

CAS 83-01-2
Molecular Formula C13H10ClNO
Molecular Weight (g/mol) 231.68
MDL Number MFCD00000633
InChI Key XNBKKRFABABBPM-UHFFFAOYSA-N
Synonym diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl
PubChem CID 65741
IUPAC Name N,N-diphenylcarbamoyl chloride
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC167140250
SDS View Documents
Thermo Scientific Chemicals
167140250
25 g Glass Bottle
Each for $127.67
Only null left
 
AC167141000
SDS View Documents
Thermo Scientific Chemicals
167141000
100 g Glass Bottle
Each for $288.76
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

83-01-2
231.68
XNBKKRFABABBPM-UHFFFAOYSA-N
65741
C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl
C13H10ClNO
MFCD00000633
diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl
N,N-diphenylcarbamoyl chloride

Specifications

81.0°C to 85.0°C
Beige to Gray-Green
97.5% min. (GC)
Glass Bottle
MFCD00000633
diphenylcarbamyl chloride, diphenylcarbamoyl chloride, carbamic chloride, diphenyl, diphenylcarbamic chloride, n,n-diphenylcarbamyl chloride, diphenylchloroformamide, carbamoyl chloride, diphenyl, carabamic chloride, diphenyl, chloroformic acid diphenylamide, carbamic chloride, n,n-diphenyl
XNBKKRFABABBPM-UHFFFAOYSA-N
N,N-diphenylcarbamoyl chloride
25 g
231.68
Crystalline Powder
83-01-2
Authentic
C13H10ClNO
(C6H5)2NCOCl
15, 3351
Solubility in water: reacts.
C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl
231.68
65741
98%
Diphenylcarbamyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES:

GHS Signal Word: Danger

EINECSNumber : 201-450-2

SDS
Documents

Documents

RUO – Research Use Only