missing translation for 'onlineSavingsMsg'
Learn More

Diphenylamine, 1% in Sulfuric Acid, Certified, LabChem™

Catalog No. LC136507
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC136507 125 mL Amber Glass
LC136501 500 mL Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC136507 Supplier LabChem Supplier No. LC136507

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5 , 122-39-4
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Max % 95.04, 3.96, 1
Color Colorless
Density 1.84g/mL
Packaging Amber Glass
Linear Formula C12H11N
Quantity 125 mL
UN Number UN1830
Solubility Information Soluble in water
Formula Weight 169.22
Grade Certified
Physical Form Liquid
Chemical Name or Material Diphenylamine, 1% in Sulfuric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause damage to organs (liver, kidneys, blood) through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature