missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylacetyl Chloride 98.0+%, TCI America™
$115.57 - $406.37
Chemical Identifiers
| CAS | 1871-76-7 |
|---|---|
| Molecular Formula | C14H11ClO |
| Molecular Weight (g/mol) | 230.691 |
| MDL Number | MFCD00013655 |
| InChI Key | MSYLETHDEIJMAF-UHFFFAOYSA-N |
| Synonym | diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride |
| PubChem CID | 74637 |
| IUPAC Name | 2,2-diphenylacetyl chloride |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl |
Chemical Identifiers
| 1871-76-7 | |
| 230.691 | |
| MSYLETHDEIJMAF-UHFFFAOYSA-N | |
| 74637 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl |
| C14H11ClO | |
| MFCD00013655 | |
| diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride | |
| 2,2-diphenylacetyl chloride |
Specifications
| 1871-76-7 | |
| White-Yellow | |
| MFCD00013655 | |
| 3261 | |
| MSYLETHDEIJMAF-UHFFFAOYSA-N | |
| 2,2-diphenylacetyl chloride | |
| 74637 | |
| ≥98.0% (GC,T) | |
| Diphenylacetyl Chloride |
| 53°C | |
| C14H11ClO | |
| 5 g | |
| diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl | |
| 230.691 | |
| 230.69 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : AO6750000
TSCA : No