Learn More
Diphenylacetyl chloride, 97.5%, Thermo Scientific Chemicals
$81.66 - $81.66
Chemical Identifiers
| CAS | 1871-76-7 |
|---|---|
| MDL Number | MFCD00013655 |
| InChI Key | MSYLETHDEIJMAF-UHFFFAOYSA-N |
| Synonym | diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride |
| PubChem CID | 74637 |
| IUPAC Name | 2,2-diphenylacetyl chloride |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC292020050
|
Thermo Scientific Chemicals
292020050 |
5 g | Glass bottle |
Each for $81.66
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1871-76-7 | |
| MSYLETHDEIJMAF-UHFFFAOYSA-N | |
| 74637 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl |
| MFCD00013655 | |
| diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride | |
| 2,2-diphenylacetyl chloride |
Specifications
| 1871-76-7 | |
| Yellow | |
| >110°C | |
| 97.5% | |
| C14H11ClO | |
| MFCD00013655 | |
| 09, 674 | |
| diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride | |
| MSYLETHDEIJMAF-UHFFFAOYSA-N | |
| 2,2-diphenylacetyl chloride | |
| 74637 | |
| 97.50% | |
| Diphenylacetyl chloride, 97.5% |
| 51°C to 56°C | |
| 175°C to 176°C (17.0mmHg) | |
| Authentic | |
| Glass bottle | |
| (C6H5)2CHCOCl | |
| 5 g | |
| 15, 3346 | |
| Solubility in water: may decompose | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl | |
| 230.69 | |
| 230.69 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 217-493-5
RTECSNumber : AO6750000
RUO â Research Use Only