Learn More
Diphenylacetyl chloride, 90+%
CAS: 1871-76-7 | C14H11ClO | 230.691 g/mol
$45.73 - $153.22
Chemical Identifiers
| CAS | 1871-76-7 |
|---|---|
| Molecular Formula | C14H11ClO |
| Molecular Weight (g/mol) | 230.691 |
| MDL Number | MFCD00013655 |
| InChI Key | MSYLETHDEIJMAF-UHFFFAOYSA-N |
| Synonym | diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride |
| PubChem CID | 74637 |
| IUPAC Name | 2,2-diphenylacetyl chloride |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0017109
|
Thermo Scientific Chemicals
L0017109 |
10 g |
Each for $45.73
|
|
|||||
|
AAL0017118
|
Thermo Scientific Chemicals
L0017118 |
50 g |
Each for $153.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1871-76-7 | |
| 230.691 | |
| MSYLETHDEIJMAF-UHFFFAOYSA-N | |
| 74637 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl |
| C14H11ClO | |
| MFCD00013655 | |
| diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride | |
| 2,2-diphenylacetyl chloride |
Specifications
| 1871-76-7 | |
| 175°C to 176°C (17 mmHg) | |
| MFCD00013655 | |
| UN3261 | |
| Moisture sensitive | |
| diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl | |
| 230.691 | |
| 230.69 | |
| Diphenylacetyl chloride |
| 50°C to 56°C | |
| C14H11ClO | |
| 10 g | |
| 1211588 | |
| 14,3316 | |
| MSYLETHDEIJMAF-UHFFFAOYSA-N | |
| 2,2-diphenylacetyl chloride | |
| 74637 | |
| ≥90% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 217-493-5
RTECSNumber : AO6750000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only