missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetyl chloride, 90+%

Catalog No. AAL0017118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0017118 50 g
AAL0017109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0017118 Supplier Thermo Scientific Chemicals Supplier No. L0017118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1871-76-7 | C14H11ClO | 230.691 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1871-76-7
Molecular Formula C14H11ClO
Molecular Weight (g/mol) 230.691
MDL Number MFCD00013655
InChI Key MSYLETHDEIJMAF-UHFFFAOYSA-N
Synonym diphenylacetyl chloride, diphenylacetic acid chloride, diphenylacetylchloride, acetyl chloride, diphenyl, unii-6553c1cb9s, alpha-phenylbenzeneacetyl chloride, diphenyl-acetyl chloride, benzeneacetyl chloride, .alpha.-phenyl, a-phenylbenzeneacetyl chloride, diphenyl acetyl chloride
PubChem CID 74637
IUPAC Name 2,2-diphenylacetyl chloride
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)Cl

Specifications

Melting Point 50°C to 56°C
Boiling Point 175°C to 176°C (17 mmHg)
Quantity 50 g
UN Number UN3261
Beilstein 1211588
Sensitivity Moisture sensitive
Merck Index 14,3316
Formula Weight 230.69
Percent Purity ≥90%
Chemical Name or Material Diphenylacetyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 217-493-5
RTECSNumber AO6750000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only