missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetonitrile 99.0+%, TCI America™

Catalog No. p-7126111
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D026125G 25 g
D0261500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D026125G Supplier TCI America Supplier No. D026125G
Only null left

Chemical Identifiers

CAS 86-29-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00001862
InChI Key NEBPTMCRLHKPOB-UHFFFAOYSA-N
Synonym diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril
PubChem CID 6837
IUPAC Name 2,2-diphenylacetonitrile
SMILES C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2

Specifications

Melting Point 75°C
Color White-Yellow
Boiling Point 181°C
Quantity 25 g
Formula Weight 193.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylacetonitrile
EINECSNumber (9)-0730
RTECSNumber AL9800000
TSCA Yes