missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetonitrile, MP Biomedicals™

Catalog No. ICN15097080
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15097080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15097080 Supplier MP Biomedicals Inc Supplier No. 0215097080

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in synthesis of methadone, antispasmodics and other pharmaceuticals

Chemical Identifiers

CAS 86-29-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
InChI Key NEBPTMCRLHKPOB-UHFFFAOYSA-N
Synonym diphenylacetonitrile, dipan, diphenatrile, benzhydrylcyanide, diphenylmethylcyanide, benzyhydrylcyanide, acetonitrile, diphenyl, benzeneacetonitrile, alpha-phenyl, usaf kf-13, difenylacetonitril
PubChem CID 6837
IUPAC Name 2,2-diphenylacetonitrile
SMILES C1=CC=C(C=C1)C(C#N)C2=CC=CC=C2

Specifications

Quantity 100 g
Formula Weight 193.2
Physical Form Powder
Chemical Name or Material Diphenylacetonitrile