missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7126476
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D086925G 25 g
D0869100G 100 g
D0869500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D086925G Supplier TCI America Supplier No. D086925G
Only null left

Chemical Identifiers

CAS 117-34-0
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00004251
InChI Key PYHXGXCGESYPCW-UHFFFAOYSA-N
Synonym diphenylacetic acid, diphenylethanoic acid, acetic acid, diphenyl, benzeneacetic acid, .alpha.-phenyl, 1,1-diphenylacetic acid, unii-658ncz0nko, diphenyl-acetic acid, alpha-toluic acid, alpha-phenyl, 658ncz0nko, benzeneacetic acid, alpha-phenyl
PubChem CID 8333
ChEBI CHEBI:41967
IUPAC Name 2,2-diphenylacetic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 146°C
Color White
Quantity 25 g
Formula Weight 212.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylacetic Acid
EINECSNumber (9)-1874
RTECSNumber AH2515000
TSCA Yes