missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phosphorochloridate, 97%

CAS: 2524-64-3 | C12H10ClO3P | 268.633 g/mol

Supplier:  Thermo Scientific Chemicals A1354618

 View more versions of this product

Catalog No. AAA1354618

Only null left
Explore available promotions

Description

Description

Diphenyl phosphorochloridate is used to prepare cyclohexyl-amidophosphoric acid diphenyl ester by reacting with cyclohexylamine. It is also used in the synthesis of alfa-substituted beta-lactams, anhydrides, esters and thioesters. It is also employed in the conversion of aldoximes to nitriles. Further, it is used in the preparation and reaction of enol phosphates, guanosine-adenosine-5',5'-triphosphate, guanosine-cytidine-5',5'-triphosphate, guanosine-uridine-5',5'-triphosphate, diguanosine-5',5'-diphosphate and diguanosine-5',5'-triphosphate. In addition to this, it serves as a phosphorylating agent for the preparation of 2-deoxy-D-galactose-3 and -6 phosphoric acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl phosphorochloridate is used to prepare cyclohexyl-amidophosphoric acid diphenyl ester by reacting with cyclohexylamine. It is also used in the synthesis of alfa-substituted beta-lactams, anhydrides, esters and thioesters. It is also employed in the conversion of aldoximes to nitriles. Further, it is used in the preparation and reaction of enol phosphates, guanosine-adenosine-5′,5′-triphosphate, guanosine-cytidine-5′,5′-triphosphate, guanosine-uridine-5′,5′-triphosphate, diguanosine-5′,5′-diphosphate and diguanosine-5′,5′-triphosphate. In addition to this, it serves as a phosphorylating agent for the preparation of 2-deoxy-D-galactose-3 and -6 phosphoric acids.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.
Specifications

Specifications

Diphenyl phosphorochloridate
1.296
113°C (235°F)
1.55
50 g
654130
diphenyl chlorophosphate, diphenyl phosphorochloridate, diphenylchlorophosphate, phosphorochloridic acid, diphenyl ester, chlorodiphenyl phosphate, o,o-diphenyl chlorophosphate, chlorodiphenoxyphosphine oxide, diphenoxychlorophosphine oxide, diphenyl chlorophosphonate, diphenylphosphoric acid monochloride
BHIIGRBMZRSDRI-UHFFFAOYSA-N
[chloro(phenoxy)phosphoryl]oxybenzene
75654
97%
2524-64-3
145°C to 147°C (1 mmHg)
C12H10ClO3P
MFCD00003030
UN3265
Moisture sensitive
Immiscible with water.
C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)Cl
268.633
268.64

Chemical Identifiers

2524-64-3
268.633
BHIIGRBMZRSDRI-UHFFFAOYSA-N
75654
C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)Cl
C12H10ClO3P
MFCD00003030
diphenyl chlorophosphate, diphenyl phosphorochloridate, diphenylchlorophosphate, phosphorochloridic acid, diphenyl ester, chlorodiphenyl phosphate, o,o-diphenyl chlorophosphate, chlorodiphenoxyphosphine oxide, diphenoxychlorophosphine oxide, diphenyl chlorophosphonate, diphenylphosphoric acid monochloride
[chloro(phenoxy)phosphoryl]oxybenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 219-759-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only