missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phosphoramidate, 97%

Catalog No. AAL0715514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0715514 25 g
AAL0715506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0715514 Supplier Thermo Scientific Chemicals Supplier No. L0715514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2015-56-7 | C12H12NO3P | 249.206 g/mol

Diphenyl phosphoramidate is used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl phosphoramidate is used as primary and secondary intermediates.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 2015-56-7
Molecular Formula C12H12NO3P
Molecular Weight (g/mol) 249.206
MDL Number MFCD00014078
InChI Key QWMUDOFWQWBHFI-UHFFFAOYSA-N
Synonym diphenyl phosphoramidate, diphenyl amidophosphate, phosphoramidic acid, diphenyl ester, diphenylphosphoramidate, amino phenoxy phosphoryl oxy benzene, diphenyl phosphoroamidate, aminodiphenoxyphosphino-1-one, amidophosphoric acid, diphenyl ester, acmc-1cki3
PubChem CID 74836
IUPAC Name [amino(phenoxy)phosphoryl]oxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(N)OC2=CC=CC=C2

Specifications

Melting Point 147°C to 151°C
Quantity 25 g
Beilstein 2054166
Solubility Information Insoluble in water.
Formula Weight 249.21
Percent Purity 97%
Chemical Name or Material Diphenyl phosphoramidate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-945-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only