missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl phosphite, contains varying amounts of phenol and (C6H5O)3P

Catalog No. p-122887
Change view
Click to view available options
Quantity:
5 g
500 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117390050 5 g Glass bottle
AC117395000 500 g Glass bottle
AC117390010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC117390050 Supplier Thermo Scientific Chemicals Supplier No. 117390050
Only null left

CAS: 4712-55-4 | C12H11O3P | 234.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4712-55-4
Molecular Formula C12H11O3P
Molecular Weight (g/mol) 234.19
MDL Number MFCD00044497
InChI Key CDXVUROVRIFQMV-UHFFFAOYSA-N
Synonym diphenyl phosphite, diphenyl phosphonate, phosphonic acid, diphenyl ester, diphenoxyphosphine oxide, diphenyl hydrogen phosphite, phosphorous acid, diphenyl ester, unii-5144js6xum, phosphonic acid diphenyl ester, phenyl phosphonate, pho 2hpo, phenyl phosphonate pho 2hpo
PubChem CID 6327546
IUPAC Name oxo(diphenoxy)phosphanium
SMILES C1=CC=C(C=C1)O[P+](=O)OC2=CC=CC=C2

Specifications

Acidity 15mg KOH/g max.
Melting Point 12°C
Density 1.2170g/mL
Boiling Point 218°C to 219°C (26.0 mmHg)
Flash Point 176°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5565 to 1.5585
Linear Formula (C6H5O)2P(O)H
Quantity 5 g
Beilstein 06, I, 94
Fieser 04,210
Specific Gravity 1.217
Solubility Information Solubility in water: insoluble
Formula Weight 234.19
Physical Form Liquid
Chemical Name or Material Diphenyl phosphite
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-202-8
TSCA TSCA

RUO – Research Use Only