accessibility menu, dialog, popup

UserName

OPOO

CAS RN 4712-55-4

IUPAC Name: oxodiphenoxy-λ⁵-phosphanylium
Synonyms: oxodiphenoxy-λ⁵-phosphanylium
Molecular Weight (g/mol): 233.18
Molecular Formula: C12H10O3P
InChi Key: CDXVUROVRIFQMV-UHFFFAOYSA-N
SMILES: O=[P+](OC1=CC=CC=C1)OC1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)