missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Phosphite (contains ca. 5% Phenol) 75.0+%, TCI America™

Catalog No. D0907500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0907500G 500 g
D090725G 25 g
D0907100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0907500G Supplier TCI America Supplier No. D0907500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4712-55-4
Molecular Formula C12H10O3P+
Molecular Weight (g/mol) 233.183
MDL Number MFCD00044497
InChI Key CDXVUROVRIFQMV-UHFFFAOYSA-N
Synonym diphenyl phosphite, diphenyl phosphonate, phosphonic acid, diphenyl ester, diphenoxyphosphine oxide, diphenyl hydrogen phosphite, phosphorous acid, diphenyl ester, unii-5144js6xum, phosphonic acid diphenyl ester, phenyl phosphonate, pho 2hpo, phenyl phosphonate pho 2hpo
PubChem CID 6327546
IUPAC Name oxo(diphenoxy)phosphanium
SMILES C1=CC=C(C=C1)O[P+](=O)OC2=CC=CC=C2

Specifications

Melting Point 12°C
Color Yellow
Boiling Point 148°C
Quantity 500 g
UN Number 1760
Formula Weight 234.19
Percent Purity ≥75.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Diphenyl Phosphite (contains ca. 5% Phenol)
EINECSNumber (3)-2602
TSCA Yes